Learn More
Enzo Life Sciences Substance P (5mg). CAS: 33507-63-0
SDP
Supplier:  Enzo Life Sciences ALX167003M005
Encompass_Preferred
Neuropeptide inducing Ca2+ mobilization. Activates G1-Proteins directly. Induces superoxide production in neutrophils. Formula: C63H98N18O13S. MW: 1347.6. Purity: ≥97% (HPLC). Formulation: Lyophilized. MI: 14: 8863. Solubility: Soluble in DMSO or water. Long Term Storage: -20°C.
Catalog No. 50-200-7868