Learn More
TARGETMOL CHEMICALS INC SUBSTANCE P 5MG
SDP
Catalog No. 502576167
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Substance P (Neurokinin P) (SP) is an undecapeptide (a peptide composed of a chain of 11 amino acid residues) member of the tachykinin neuropeptide family. purity: 98%
Catalog No. 50-257-6167 Supplier TARGETMOL CHEMICALS INC Supplier No. TP10875MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Substance P (Neurokinin P) (SP) is an undecapeptide (a peptide composed of a chain of 11 amino acid residues) member of the tachykinin neuropeptide family. purity: 98%