Learn More
Medchemexpress LLC Substance P (7-11) | 51165-05-0 | 99.1% | 612.78 | 5 MG
SDP
Catalog No. 5000423087
Encompass_Preferred
Substance P (7-11) is a C-terminal fragment of Substance P. It is for research use only and can cause an increase in the intracellular calcium concentration. In vitro, it also increases PGE2 and collagenase production at concentrations greater than 1 μM.
  • Increases PGE2 and collagenase production at concentrations greater than 1 μM
  • Increases intracellular calcium concentration as measured by the fluorescent dye Fura-2
  • Maximal change in intracellular calcium induced by 10 μM was 140±30 nM
Catalog No. 50-004-23087 Supplier Medchemexpress LLC Supplier No. HYP14925MG
Substance P (7-11) is a C-terminal fragment of Substance P. It is for research use only and can cause an increase in the intracellular calcium concentration. In vitro, it also increases PGE2 and collagenase production at concentrations greater than 1 μM.
  • Increases PGE2 and collagenase production at concentrations greater than 1 μM
  • Increases intracellular calcium concentration as measured by the fluorescent dye Fura-2
  • Maximal change in intracellular calcium induced by 10 μM was 140±30 nM