Learn More
Medchemexpress LLC Substance P, FAM-labeled (TFA) | 98.3% | 1705.93 | 1 MG
SDP
Catalog No. 5000426720
Encompass_Preferred
Substance P, FAM-labeled (TFA) is a neuropeptide that acts as a neurotransmitter and neuromodulator in the central nervous system. It is labeled with a FAM fluorophore and is suitable for research applications.
  • Labeled with FAM fluorophore
  • Acts as a neurotransmitter and neuromodulator
  • Endogenous receptor is neurokinin 1 receptor (NK1R)
  • Suitable for research use only
  • High purity: 98.3%
  • Available as a solid, light yellow to yellow appearance
  • Soluble in DMSO (≥ 100 mg/mL)
Catalog No. 50-004-26720 Supplier Medchemexpress LLC Supplier No. HYP0201AF1MG
Substance P, FAM-labeled (TFA) is a neuropeptide that acts as a neurotransmitter and neuromodulator in the central nervous system. It is labeled with a FAM fluorophore and is suitable for research applications.
  • Labeled with FAM fluorophore
  • Acts as a neurotransmitter and neuromodulator
  • Endogenous receptor is neurokinin 1 receptor (NK1R)
  • Suitable for research use only
  • High purity: 98.3%
  • Available as a solid, light yellow to yellow appearance
  • Soluble in DMSO (≥ 100 mg/mL)