Learn More
Medchemexpress LLC Substance P (Neurokinin P) | 33507-63-0 | 99.5% | 1347.63 | 25 MG
SDP
Catalog No. 5000426145
Encompass_Preferred
Substance P (Neurokinin P) is a CNS-penetrant neuropeptide that functions as both a neurotransmitter and a neuromodulator within the central nervous system. Its endogenous receptor is the neurokinin 1 receptor (NK1-receptor, NK1R). This compound is intended for research use only.
  • Acts as a CNS-penetrant neuropeptide.
  • Functions as a neurotransmitter and neuromodulator.
  • Endogenous receptor is the neurokinin 1 receptor (NK1R).
  • Induces clathrin-dependent internalization of the NK1-R in transfected epithelial cells.
  • Substance P/NK1-R complex dissociates in acidified endosomes, where Substance P is degraded, and the NK1-R recycles to the cell surface.
Catalog No. 50-004-26145 Supplier Medchemexpress LLC Supplier No. 25MGHYP0201
Substance P (Neurokinin P) is a CNS-penetrant neuropeptide that functions as both a neurotransmitter and a neuromodulator within the central nervous system. Its endogenous receptor is the neurokinin 1 receptor (NK1-receptor, NK1R). This compound is intended for research use only.
  • Acts as a CNS-penetrant neuropeptide.
  • Functions as a neurotransmitter and neuromodulator.
  • Endogenous receptor is the neurokinin 1 receptor (NK1R).
  • Induces clathrin-dependent internalization of the NK1-R in transfected epithelial cells.
  • Substance P/NK1-R complex dissociates in acidified endosomes, where Substance P is degraded, and the NK1-R recycles to the cell surface.