Learn More
Biomatik Corporation Substance P (SP) ELISA Kit
Supplier:  Biomatik Corporation EKU12337
Encompass_Preferred
Substance P (SP) ELISA Kit for detecting SP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.63pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6423