Learn More
Medchemexpress LLC Substance P TFA (neurokinin P TFA) | 148470-19-3 | 99.3% | 1461.65 | 25 MG
SDP
Catalog No. 5000425831
Encompass_Preferred
Substance P TFA (neurokinin P TFA) is a CNS-penetrant neuropeptide that functions as a neurotransmitter and neuromodulator within the central nervous system. Its endogenous receptor is the neurokinin 1 receptor (NK1-receptor, NK1R), playing a crucial role in various biological activities.
  • Acts as a CNS-penetrant neuropeptide
  • Functions as a neurotransmitter and neuromodulator
  • Endogenous receptor is neurokinin 1 receptor (NK1R)
  • Appears as a white to off-white solid
  • Soluble in H2O and PBS with ultrasonic assistance
  • Involved in the internalization and recycling of the NK1R complex
  • Related to peptides, hormones, and neuropeptides classifications
  • Associated with research in cancer, neurological, and metabolic diseases
Catalog No. 50-004-25831 Supplier Medchemexpress LLC Supplier No. 25MGHYP0201A
Substance P TFA (neurokinin P TFA) is a CNS-penetrant neuropeptide that functions as a neurotransmitter and neuromodulator within the central nervous system. Its endogenous receptor is the neurokinin 1 receptor (NK1-receptor, NK1R), playing a crucial role in various biological activities.
  • Acts as a CNS-penetrant neuropeptide
  • Functions as a neurotransmitter and neuromodulator
  • Endogenous receptor is neurokinin 1 receptor (NK1R)
  • Appears as a white to off-white solid
  • Soluble in H2O and PBS with ultrasonic assistance
  • Involved in the internalization and recycling of the NK1R complex
  • Related to peptides, hormones, and neuropeptides classifications
  • Associated with research in cancer, neurological, and metabolic diseases