Learn More
Pickering Laboratories Inc Substitute Ocean Water, ASTM D1141-98 (2003), Not Stabilized, case of 4 (950mL/bottle)
SDP
Catalog No. 501974241
Encompass_Preferred
Substitute Ocean Water is prepared according to official ASTM method D1141-98. This product could be successfully used in a wide variety of tests where solution simulating sea water is required, such as oil contamination testing, detergent evaluation and corrosion testing. The pH of the solution is 8.2. The solution is ready to use and is stored at room temperature. Inquire about different sizes.
Catalog No. 50-197-4241 Supplier Pickering Laboratories Inc Supplier No. 17000801
Add to Cart
Add to Cart
Substitute Ocean Water is prepared according to official ASTM method D1141-98. This product could be successfully used in a wide variety of tests where solution simulating sea water is required, such as oil contamination testing, detergent evaluation and corrosion testing. The pH of the solution is 8.2. The solution is ready to use and is stored at room temperature. Inquire about different sizes.