Learn More
Cayman Chemical Substance P (trifluoroacetate salt), Purity: ≥95%, CAS: 148470-19-3, 10 mg
SDP
Catalog No. 502933918
Encompass_Preferred
An endogenous NK1 receptor agonist (Ki = 0.14 and 300 nM for NK1 and NK3, respectively); ubiquitously expressed in the body; released in the dorsal horn of the spinal cord where it induces pain through NK1 receptors; involved in depression, anxiety, seizures, and emesis among other functions
Catalog No. 50-293-3918 Supplier Cayman Chemical Supplier No. 2403510 MG
An endogenous NK1 receptor agonist (Ki = 0.14 and 300 nM for NK1 and NK3, respectively); ubiquitously expressed in the body; released in the dorsal horn of the spinal cord where it induces pain through NK1 receptors; involved in depression, anxiety, seizures, and emesis among other functions