Learn More
MOLECULAR DEPOT LLC Substrate Buffer - 50 mL
Catalog No. 5000013510
Encompass_Preferred
Beta-Galactosidase Chromogenic Substrate Buffer is a highly optimized buffer for the specific stabilization of beta-galactosidase chromogenic substrates This product has been used as stabilizing agent in various beta-galactosidase-related applications For in vitro research use only
Catalog No. 50-000-13510 Supplier MOLECULAR DEPOT LLC Supplier No. B2010650
Beta-Galactosidase Chromogenic Substrate Buffer is a highly optimized buffer for the specific stabilization of beta-galactosidase chromogenic substrates This product has been used as stabilizing agent in various beta-galactosidase-related applications For in vitro research use only