Learn More
Electron Microscopy Sciences EMS Substrate Gold Coated Round Coverslips, 16 mm
SDP
Catalog No. 501929310
Encompass_Preferred
EMS offers a variety of economically priced coated substrates. All coatings are deposited under vacuum using electron beam evaporation. These high quality substrates are coated with 500 Angstroms (50 nm) of gold, with a thin chromium adhesion layer (2-7 nm) between the glass and coating. All substrates are pre-cleaned using an ion source for quality assurance. Autoclavable. Packed in a cleanroom. Shipped in X0 Gel-Pak.
Dimensions: 16 mm
Material: Borosilicate glass
Catalog No. 50-192-9310 Supplier Electron Microscopy Sciences Supplier No. 7189225
EMS offers a variety of economically priced coated substrates. All coatings are deposited under vacuum using electron beam evaporation. These high quality substrates are coated with 500 Angstroms (50 nm) of gold, with a thin chromium adhesion layer (2-7 nm) between the glass and coating. All substrates are pre-cleaned using an ion source for quality assurance. Autoclavable. Packed in a cleanroom. Shipped in X0 Gel-Pak.
Dimensions: 16 mm
Material: Borosilicate glass