Learn More
Bachem Ac-Tyr-OEt
SDP
Supplier:  Bachem 4000673.0025
Encompass_Preferred
Ac-Tyr-OEt, 840-97-1, C13H17NO4, Mr 251.28, 25g. L-ATEE. Substrate for chymotrypsin and carboxypeptidase Y.
Catalog No. 50-194-7015