Learn More
Bachem H-Gly-D-Phe-OH
SDP
Supplier:  Bachem 4006237.0005
Encompass_Preferred
H-Gly-D-Phe-OH, 34258-14-5, C11H14N2O3, Mr 222.24, 5g. Substrate for D-peptidase S, an intracellular carboxypeptidase-like enzyme that preferentially hydrolyzes dipeptides containing hydrophobic D-amino acids.
Catalog No. 50-194-6831