Learn More
Bachem FA-Ala-Arg-OH
SDP
Supplier:  Bachem 4013709.0250
Encompass_Preferred
FA-Ala-Arg-OH, 76079-06-6, C16H23N5O5, Mr 365.39, 250mg. Substrate for human plasma carboxypeptidase N and membrane-bound carboxypeptidase D.
Catalog No. 50-194-7053