Learn More
Bachem H-Hyp-Gly-OH
SDP
Supplier:  Bachem 4001823.0001
Encompass_Preferred
H-Hyp-Gly-OH, 7672-35-7, C7H12N2O4, Mr 188.18, 1g. Substrate for prolinase (prolyl dipeptidase). Inoue et al. showed in a clinical study that ingestion of the dipeptides Hyp-Gly and Pro-Hyp could improve facial skin moisture and elasticity and reduce facial ageing.
Catalog No. 50-194-6832