Learn More
Bachem Pyr-Val-OH
SDP
Supplier:  Bachem 4001328.0001
Encompass_Preferred
Pyr-Val-OH , Pyr-Val-OH, 21282-10-0, C10H16N2O4, Mr 228.25, 1g. Substrate for pyroglutamyl peptidase I.
Catalog No. 50-194-6859