Learn More
Bachem Pyr-bNA
SDP
Catalog No. 501947074
Encompass_Preferred
Pyr-bNA, 22155-91-5, C15H14N2O2, Mr 254.28, 5g. Substrate for the assay of pyroglutamyl peptidase I.
Catalog No. 50-194-7074 Supplier Bachem Supplier No. 4003907.5000
Pyr-bNA, 22155-91-5, C15H14N2O2, Mr 254.28, 5g. Substrate for the assay of pyroglutamyl peptidase I.