Learn More
Bachem 4-Nitrophenyl b-D-galactopyranoside
SDP
Supplier:  Bachem 4013562.0025
Encompass_Preferred
4-Nitrophenyl b-D-galactopyranoside, 3150-24-1, C12H15NO8, Mr 301.25, 25g. 4-Nitrophenyl b-D-galactoside-(1,5). Substrate for the determination of b-galactosidase.
Catalog No. 50-194-7042