Learn More
Bachem Z-Arg-pNA · HCl
SDP
Catalog No. 501947079
Encompass_Preferred
Z-Arg-pNA · HCl, 59188-53-3, C20H24N6O5 · HCl, Mr 464.91, 5g. L-ZAPA. Substrate for trypsin.
Catalog No. 50-194-7079 Supplier Bachem Supplier No. 4000780.0005
Z-Arg-pNA · HCl, 59188-53-3, C20H24N6O5 · HCl, Mr 464.91, 5g. L-ZAPA. Substrate for trypsin.