Learn More
CPC Scientific H-Arg-Arg-Leu-Ile-Glu-Asp-Asn-Glu-Tyr-Thr-Ala-Arg-Gly-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PKCP049A
Encompass
ONE-LETTER SEQUENCE: RRLIEDNEYTARGMOLECULAR FORMULA: C66H109N23O23MOLECULAR WEIGHT:1592.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [81493-98-3], RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: E.G. Krebs et al., Proc. Nat. Acad. Sci. USA, 79, 1443 (1982)
Catalog No. 50-211-0203