Learn More
Vector Laboratories VECTOR Red Alkaline Phosphatase (AP) Substrate Kit 200 ml
SDP
Catalog No. NC9250081
Encompass
VECTOR® Red AP Substrate Kit produces a magenta heat-stable reaction product that is also fluorescent. Sections stained with the Vector Red substrate can be dehydrated, cleared, and permanently mounted. This chromogen can be used singly or in combination with other alkaline phosphatase or peroxidase substrates for multiple label applications. Because of its stability under ISH conditions, VECTOR Red can be used in IHC/ISH double labeling applications. The bright red to orange fluorescence of the Vector Red reaction product can be visualized using DyLight® 594, Texas Red® or TRITC filter systems. This substrate can also be used for ISH or on blots. This kit contains stock solutions in convenient dropper bottles, sufficient for 200 ml working solution.
Catalog No. NC9250081 Supplier Vector Laboratories Supplier No. SK5100
VECTOR® Red AP Substrate Kit produces a magenta heat-stable reaction product that is also fluorescent. Sections stained with the Vector Red substrate can be dehydrated, cleared, and permanently mounted. This chromogen can be used singly or in combination with other alkaline phosphatase or peroxidase substrates for multiple label applications. Because of its stability under ISH conditions, VECTOR Red can be used in IHC/ISH double labeling applications. The bright red to orange fluorescence of the Vector Red reaction product can be visualized using DyLight® 594, Texas Red® or TRITC filter systems. This substrate can also be used for ISH or on blots. This kit contains stock solutions in convenient dropper bottles, sufficient for 200 ml working solution.