Learn More
Diapharma Group, Inc. SUBSTRATE S-2366(25MGVIAL
SDP
Catalog No. NC9817128
Encompass
Chromogenic substrate for activated protein C and factor XIa Package: 25 mg Molecular wt: 539.0 Formula: pyroGlu-Pro-Arg-pNA -- HCl Solubility: >10mmol/L in H 2 O
Catalog No. NC9817128 Supplier Diapharma Group, Inc. Supplier No. S821090
Add to Cart
Add to Cart
Chromogenic substrate for activated protein C and factor XIa Package: 25 mg Molecular wt: 539.0 Formula: pyroGlu-Pro-Arg-pNA -- HCl Solubility: >10mmol/L in H 2 O