Learn More
Electron Microscopy Sciences Substrate of Silver Coated 4" Silicon Wafer, P-Type <100>, 100 mm
SDP
, 100 mm">
Catalog No. 501929860
Encompass_Preferred
EMS offers a variety of economically priced coated substrates. All coatings are deposited under vacuum using electron beam evaporation. These high quality substrates are coated with 500 Angstroms (50 nm) of silver, with a thin titanium adhesion layer (2-7 nm) between the glass and silver coating. All substrates are pre-cleaned using an ion source for quality assurance. Autoclavable. P-Type <100> wafer.
Dimensions: 100 mm
Material: Silicon Wafer P/Boron
Catalog No. 50-192-9860 Supplier Electron Microscopy Sciences Supplier No. 7189235
EMS offers a variety of economically priced coated substrates. All coatings are deposited under vacuum using electron beam evaporation. These high quality substrates are coated with 500 Angstroms (50 nm) of silver, with a thin titanium adhesion layer (2-7 nm) between the glass and silver coating. All substrates are pre-cleaned using an ion source for quality assurance. Autoclavable. P-Type <100> wafer.
Dimensions: 100 mm
Material: Silicon Wafer P/Boron