Learn More
Electron Microscopy Sciences EMS Substrate Gold Coated 2'' Silicon Wafer, P-Type <100>, 50 mm
SDP
Catalog No. 501929308
Encompass_Preferred
EMS offers a variety of economically priced coated substrates. All coatings are deposited under vacuum using electron beam evaporation. These high quality substrates are coated with 500 Angstroms (50 nm) of gold, with a thin chromium adhesion layer (2-7 nm) between the glass and coating. All substrates are pre-cleaned using an ion source for quality assurance. Autoclavable.
Dimensions: 50 mm
Material: Silicon Wafer P/Boron
Catalog No. 50-192-9308 Supplier Electron Microscopy Sciences Supplier No. 7189215
EMS offers a variety of economically priced coated substrates. All coatings are deposited under vacuum using electron beam evaporation. These high quality substrates are coated with 500 Angstroms (50 nm) of gold, with a thin chromium adhesion layer (2-7 nm) between the glass and coating. All substrates are pre-cleaned using an ion source for quality assurance. Autoclavable.
Dimensions: 50 mm
Material: Silicon Wafer P/Boron