Learn More
Medchemexpress LLC Subtilisin (EC 3.4.21.14) | 9014-01-1 | 200 MG
SDP
Catalog No. 5000400351
Encompass_Preferred
Subtilisin (EC 3.4.21.14) is a bacterial serine protease produced through deep fermentation, extraction, and purification of *Bacillus subtilis*. It hydrolyzes protein molecules into polypeptides or amino acids and can catalyze polypeptide synthesis in organic solvents. This product contains stabilizers and has observed anticancer and antifouling activities.
  • Induces apoptosis
  • Stimulates expression of pro-allergic cytokines (IL-1α, IL-33)
  • Induces prototypic allergic lung inflammation
  • Exhibits anticancer activity against breast and colon cancer
  • Shows antifouling activity
  • Can be used as a detergent additive
  • Effective temperature range: 20-60°C (optimum 35-45°C)
  • Effective pH range: 6.0-11.0 (optimum 9.5-10.5)
  • Activators: Ca2+, Mg2+, Mn2+, Zn2+, Fe2+
  • Inhibitors: Reagents acting on serine such as sulfonyl halides
Catalog No. 50-004-00351 Supplier Medchemexpress LLC Supplier No. HYP2729200MG
Subtilisin (EC 3.4.21.14) is a bacterial serine protease produced through deep fermentation, extraction, and purification of *Bacillus subtilis*. It hydrolyzes protein molecules into polypeptides or amino acids and can catalyze polypeptide synthesis in organic solvents. This product contains stabilizers and has observed anticancer and antifouling activities.
  • Induces apoptosis
  • Stimulates expression of pro-allergic cytokines (IL-1α, IL-33)
  • Induces prototypic allergic lung inflammation
  • Exhibits anticancer activity against breast and colon cancer
  • Shows antifouling activity
  • Can be used as a detergent additive
  • Effective temperature range: 20-60°C (optimum 35-45°C)
  • Effective pH range: 6.0-11.0 (optimum 9.5-10.5)
  • Activators: Ca2+, Mg2+, Mn2+, Zn2+, Fe2+
  • Inhibitors: Reagents acting on serine such as sulfonyl halides