Learn More
MSC Suburban Tool Four Face Parallel, 8 x 1 x 3/4 in, Steel
Catalog No. 5000537112
Encompass
This steel four face parallel is designed for precision applications. It is sold individually and features a parallelism of 0.0001 in per 6 in.
  • Four face parallel type
  • 8 in length
  • 1 in height
  • 3/4 in thickness
  • Constructed from steel
  • 4 precision-ground sides
  • Opposite sides parallel
  • Adjacent sides square
  • Squareness of 0.0005 in per 1 in
  • Accuracy of 0.000250 in
Catalog No. 50-005-37112 Supplier MSC Supplier No. 06821268
This steel four face parallel is designed for precision applications. It is sold individually and features a parallelism of 0.0001 in per 6 in.
  • Four face parallel type
  • 8 in length
  • 1 in height
  • 3/4 in thickness
  • Constructed from steel
  • 4 precision-ground sides
  • Opposite sides parallel
  • Adjacent sides square
  • Squareness of 0.0005 in per 1 in
  • Accuracy of 0.000250 in