Learn More
MSC Suburban Tool Four-Way Steel Parallel, 6 in, 0.0001 in Parallelism, Steel
Catalog No. 5000529068
Encompass
This four-way steel parallel is engineered for high-precision measurement and setup tasks. Constructed from hardened steel, it is meticulously ground to close tolerances on all four sides, offering exceptional accuracy and flexibility for individual use or in conjunction with other parallels.
  • Made from hardened steel.
  • Ground to close tolerances on four sides.
  • Can be used individually or matched with any other individual four-way parallel.
  • Different sizes can be used together.
  • Held to size ±0.00025 in on four sides.
  • Adjacent sides square within 0.0005 in per "
  • Opposite sides parallel within 0.0001 in per 6 inches.
  • Proudly made in the U.S.A.
Catalog No. 50-005-29068 Supplier MSC Supplier No. 06821185
This four-way steel parallel is engineered for high-precision measurement and setup tasks. Constructed from hardened steel, it is meticulously ground to close tolerances on all four sides, offering exceptional accuracy and flexibility for individual use or in conjunction with other parallels.
  • Made from hardened steel.
  • Ground to close tolerances on four sides.
  • Can be used individually or matched with any other individual four-way parallel.
  • Different sizes can be used together.
  • Held to size ±0.00025 in on four sides.
  • Adjacent sides square within 0.0005 in per "
  • Opposite sides parallel within 0.0001 in per 6 inches.
  • Proudly made in the U.S.A.