Learn More
MSC Suburban Tool Magnetic Cylinder Square, 0.0001 in Accuracy, 1 in Cylinder Dia
Catalog No. 5000528144
Encompass
This magnetic cylinder square is designed to easily check squareness on component parts. Manufactured from steel and featuring a magnetic base, it offers 0.0001 inch accuracy for precise measurements while eliminating clamping distortion and speeding up setup.
  • Base is perpendicular to shaft within 0.0001 inch
  • Magnetic base eliminates clamping distortion and speeds setup
  • Manufactured from normalized steel for stability and uniformity
  • Includes fitted wooden storage case
  • Made in the U.S.A.
Catalog No. 50-005-28144 Supplier MSC Supplier No. 32147399
This magnetic cylinder square is designed to easily check squareness on component parts. Manufactured from steel and featuring a magnetic base, it offers 0.0001 inch accuracy for precise measurements while eliminating clamping distortion and speeding up setup.
  • Base is perpendicular to shaft within 0.0001 inch
  • Magnetic base eliminates clamping distortion and speeds setup
  • Manufactured from normalized steel for stability and uniformity
  • Includes fitted wooden storage case
  • Made in the U.S.A.