Learn More
Medchemexpress LLC Suc-AAPF-AMC | 88467-45-2 | MFCD00080246 | 99.5% | 661.70 | C34H39N5O9 | 5 MG
SDP
Catalog No. 5000222864
Encompass_Preferred
Suc-AAPF-AMC is a fluorogenic peptide substrate used to quantify α-chymotrypsin activity. The succinylated tetrapeptide releases 7-amino-4-methylcoumarin (AMC) upon proteolytic cleavage, producing a fluorescent signal suitable for kinetic or end-point assays. The product is supplied with purity data, a product data sheet, and a safety data sheet for handling and storage.
  • Fluorogenic substrate for α-chymotrypsin activity assays
  • AMC release produces a measurable fluorescent signal for kinetic and end-point assays
  • Available as a solid and as a 10 mM solution in DMSO for assay flexibility
  • High purity suitable for research applications
  • Includes datasheet and safety data sheet for use and storage guidance
Catalog No. 50-002-22864 Supplier Medchemexpress LLC Supplier No. HY1378115MG
Suc-AAPF-AMC is a fluorogenic peptide substrate used to quantify α-chymotrypsin activity. The succinylated tetrapeptide releases 7-amino-4-methylcoumarin (AMC) upon proteolytic cleavage, producing a fluorescent signal suitable for kinetic or end-point assays. The product is supplied with purity data, a product data sheet, and a safety data sheet for handling and storage.
  • Fluorogenic substrate for α-chymotrypsin activity assays
  • AMC release produces a measurable fluorescent signal for kinetic and end-point assays
  • Available as a solid and as a 10 mM solution in DMSO for assay flexibility
  • High purity suitable for research applications
  • Includes datasheet and safety data sheet for use and storage guidance