Learn More
Enzo Life Sciences Suc-AAPF-pNA (25mg)
SDP
Catalog No. 502011252
Encompass_Preferred
Chromogenic substrate cleaved by cathepsin G, subtilisins, chymotrypsin, chymase, and cyclophilins, but not neutrophil elastase. Release of pNA is monitored at 405-410 nm. This substrate is useful for inhibitor screening and kinetic analysis. Also available: calibration standard pNA (BML-KI145). Alternative name: Suc-Ala-Ala-Pro-Phe-pNA. Purity: ≥99%. Long Term Storage: -20°C.
Catalog No. 50-201-1252 Supplier Enzo Life Sciences Supplier No. BMLP1410025
Chromogenic substrate cleaved by cathepsin G, subtilisins, chymotrypsin, chymase, and cyclophilins, but not neutrophil elastase. Release of pNA is monitored at 405-410 nm. This substrate is useful for inhibitor screening and kinetic analysis. Also available: calibration standard pNA (BML-KI145). Alternative name: Suc-Ala-Ala-Pro-Phe-pNA. Purity: ≥99%. Long Term Storage: -20°C.