Learn More
Medchemexpress LLC Suc-AAPF-pNA (Suc-Ala-Ala-Pro-Phe-pNA) | 70967-97-4 | 99.42% | 624.64 | 25 MG
SDP
Catalog No. 5000426804
Encompass_Preferred
Suc-AAPF-pNA is a chromogenic p-nitroanilide (pNA) substrate with a Km of 1.7 mM. When cleaved, it releases 4-nitroaniline, which is yellow and can be measured spectrophotometrically. This substrate is used for measuring free and membrane-bound cathepsin G in human neutrophils.
  • Measures elastase activity
  • Used to measure cercarial elastase activity by monitoring absorbance at 400 nm
  • Hydrolysis allows for the measurement of proteolytic activity
  • Enzyme activity can be measured spectrophotometrically at 410 nm
Catalog No. 50-004-26804 Supplier Medchemexpress LLC Supplier No. 25MGHYP2573
Suc-AAPF-pNA is a chromogenic p-nitroanilide (pNA) substrate with a Km of 1.7 mM. When cleaved, it releases 4-nitroaniline, which is yellow and can be measured spectrophotometrically. This substrate is used for measuring free and membrane-bound cathepsin G in human neutrophils.
  • Measures elastase activity
  • Used to measure cercarial elastase activity by monitoring absorbance at 400 nm
  • Hydrolysis allows for the measurement of proteolytic activity
  • Enzyme activity can be measured spectrophotometrically at 410 nm