Learn More
Medchemexpress LLC Suc-AAPK-pNA | 108929-39-1 | 98.1% | 605.64 | 10 MG
SDP
Catalog No. 5000397936
Encompass_Preferred
Suc-AAPK-pNA is a chromogenic substrate used for determining serine/threonine kinase activity and enzyme kinetic parameters. This product is for research use only.
  • Chromogenic substrate
  • Used for determining serine/threonine kinase activity
  • Used for determining enzyme kinetic parameters
  • Intended for research use only
Catalog No. 50-003-97936 Supplier Medchemexpress LLC Supplier No. HYP448410MG
Suc-AAPK-pNA is a chromogenic substrate used for determining serine/threonine kinase activity and enzyme kinetic parameters. This product is for research use only.
  • Chromogenic substrate
  • Used for determining serine/threonine kinase activity
  • Used for determining enzyme kinetic parameters
  • Intended for research use only