Learn More
Bachem Suc-Ala-Ala-Val-Ala-pNA
SDP
Supplier:  Bachem 4006610.0250
Encompass_Preferred
Suc-Ala-Ala-Val-Ala-pNA, 108322-03-8, C24H34N6O9, Mr 550.57, 250mg. Suc-AAVA-pNA has been used for assaying hamster chymase 2.
Catalog No. 50-194-7026