Learn More
Cayman Chemical Suc-Ala-Ala-Ala-pNA, Purity: ≥98%, CAS: 52299-14-6, 10 mg
SDP
Catalog No. 502912510
Encompass_Preferred
A colorimetric elastase substrate; hydrolysis releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of elastase activity
Catalog No. 50-291-2510 Supplier Cayman Chemical Supplier No. 3647810 MG
A colorimetric elastase substrate; hydrolysis releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of elastase activity