Learn More
Carbosynth LLC. Suc-Ala-Ala-Ala-pNA, 0.1g, MW: 451.43, Molecular formula: C19H25N5O8, CAS# 52299-14-6
SDP
Supplier:  Carbosynth LLC. SAA30710.1G
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: [STANA] Succinyl-L-Alanyl-L-Alanyl-L-Alanine-p-Nitroanilide, Purity: , Grade AA, Application: Substrate for Elastase, Storage Temperature: 2-10C, References: J. Bieth, B. Spiess, and C.G. Wermuth, Biochem. Med., 11, 350 (1974).
Catalog No. 50-168-7005