Learn More
Bachem Suc-Ala-Ala-Phe-AMC
SDP
Supplier:  Bachem 4006306.0050
Encompass_Preferred
Suc-Ala-Ala-Phe-AMC, 71973-79-0, C29H32N4O8, Mr 564.60, 50mg. Suc-AAF-AMC, a highly sensitive fluorogenic chymotrypsin substrate. It is also hydrolyzed by neutral metalloendopeptidases, such as thermolysin and enkephalinase (neprilysin, NEP 24.11). Suc-AAF-MCA has also been used for the chymotrypsin-like activity of the 20S proteasome.
Catalog No. 50-260-303