Learn More
Bachem Suc-Ala-Ala-Pro-Glu-pNA
SDP
Catalog No. 50260423
Encompass_Preferred
Suc-Ala-Ala-Pro-Glu-pNA, 108929-37-9, C26H34N6O11, Mr 606.59, 250mg. Suc-AAPE-pNA, a substrate for a glutamyl endopeptidase, that cleaves after negatively charged residues.
Catalog No. 50-260-423 Supplier Bachem Supplier No. 4017343.0250
Add to Cart
Add to Cart
Suc-Ala-Ala-Pro-Glu-pNA, 108929-37-9, C26H34N6O11, Mr 606.59, 250mg. Suc-AAPE-pNA, a substrate for a glutamyl endopeptidase, that cleaves after negatively charged residues.