Learn More
Carbosynth LLC. Suc-Ala-Ala-Pro-Gly-pNA, 25mg, MW: (M.W. 534.53), Molecular formula: C23H30N6O9
SDP
Supplier:  Carbosynth LLC. SAP3666PI25MG
Encompass_Preferred
Application: Substrate for Pancreatic Elastase, Storage Temperature: -20°C, References: C.L. Largman, Biochemistry, 22, 3763 (1983)., E.G. Del Mar, C. Largman, J.W. Brodick, M. Fassett, and M.C. Geokas, Biochemistry, 19, 468 (1980).
Catalog No. 50-168-7030