Learn More
Bachem Suc-Ala-Ala-Pro-Phe-AMC
SDP
Catalog No. 50260318
Encompass_Preferred
Suc-Ala-Ala-Pro-Phe-AMC, 88467-45-2, C34H39N5O9, Mr 661.71, 50mg. The peptidylprolyl isomerase substrate Suc-AAPF-AMC is also hydrolyzed by carboxypeptidase Y, cathepsin G, and chymotrypsin. Suc-AAPF-AMC has also been used to analyze the chymotrypsin-like activity of trypsins.
Catalog No. 50-260-318 Supplier Bachem Supplier No. 4012873.0050
Suc-Ala-Ala-Pro-Phe-AMC, 88467-45-2, C34H39N5O9, Mr 661.71, 50mg. The peptidylprolyl isomerase substrate Suc-AAPF-AMC is also hydrolyzed by carboxypeptidase Y, cathepsin G, and chymotrypsin. Suc-AAPF-AMC has also been used to analyze the chymotrypsin-like activity of trypsins.