Learn More
Bachem Suc-Ala-Ala-Pro-Phe-pNA
SDP
Catalog No. NC9865712
Encompass_Preferred
Suc-Ala-Ala-Pro-Phe-pNA, 70967-97-4, C30H36N6O9, Mr 624.65, 50mg. SAAPNA. A readily soluble, specific and sensitive substrate for chymotrypsin and human pancreatic elastase. It is also hydrolyzed by cathepsin G and chymase. Furthermore it is the standard substrate for FK-506 binding proteins (FKBPs, also called macrophilins) and cyclophilins, which belong to the group of peptidyl prolyl cis-trans isomerases (PPIases). Thus, Suc-AAPF-pNA has been used for an uncoupled protease-free assay of PPIase activity.
Catalog No. NC9865712 Supplier Bachem Supplier No. 4002299.0050
Suc-Ala-Ala-Pro-Phe-pNA, 70967-97-4, C30H36N6O9, Mr 624.65, 50mg. SAAPNA. A readily soluble, specific and sensitive substrate for chymotrypsin and human pancreatic elastase. It is also hydrolyzed by cathepsin G and chymase. Furthermore it is the standard substrate for FK-506 binding proteins (FKBPs, also called macrophilins) and cyclophilins, which belong to the group of peptidyl prolyl cis-trans isomerases (PPIases). Thus, Suc-AAPF-pNA has been used for an uncoupled protease-free assay of PPIase activity.