Learn More
Carbosynth LLC. Suc-Ala-Leu-Pro-Phe-pNA (10mg vial), 1ea, MW: 666.72, Molecular formula: C33H42N6O9, CAS# 128802-78-8
SDP
Catalog No. 501687015
Encompass_Preferred
Synonyms: Succinyl-L-Alanyl-L-Leucyl-L-Prolyl-L-Phenylalanine p-Nitroanilide, Application: Substrate for PPlase (Peptidyl-prolyl cis-trans Isomerase), Storage Temperature: -20°C
Catalog No. 50-168-7015 Supplier Carbosynth LLC. Supplier No. SAF3162V10MG
Add to Cart
Add to Cart
Synonyms: Succinyl-L-Alanyl-L-Leucyl-L-Prolyl-L-Phenylalanine p-Nitroanilide, Application: Substrate for PPlase (Peptidyl-prolyl cis-trans Isomerase), Storage Temperature: -20°C