Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Suc-Ala-Phe-Lys-AMC | 73207-91-7 | 99.8% | 621.68 | 1 MG
SDP

Catalog No. 5000398800
Encompass_Preferred

Suc-Ala-Phe-Lys-AMC is a highly sensitive fluorogenic substrate for plasmin and is also used for assaying gingipain K.

  • Highly sensitive fluorogenic substrate for plasmin
  • Used for assaying gingipain K
  • Molecular formula: C32H39N5O8
  • Molecular weight: 621.68
  • Sequence: {Suc-Ala}-Phe-{Lys-AMC}
  • Appearance: White to off-white solid
  • Purity: 99.75%
  • Storage (powder): -80°C for 2 years; -20°C for 1 year
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light)
  • Soluble in DMSO: 100 mg/mL

Catalog No. 50-003-98800 Supplier Medchemexpress LLC Supplier No. HYP43601MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suc-Ala-Phe-Lys-AMC is a highly sensitive fluorogenic substrate for plasmin and is also used for assaying gingipain K.

  • Highly sensitive fluorogenic substrate for plasmin
  • Used for assaying gingipain K
  • Molecular formula: C32H39N5O8
  • Molecular weight: 621.68
  • Sequence: {Suc-Ala}-Phe-{Lys-AMC}
  • Appearance: White to off-white solid
  • Purity: 99.75%
  • Storage (powder): -80°C for 2 years; -20°C for 1 year
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light)
  • Soluble in DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.