Learn More
Medchemexpress LLC Suc-Ala-Phe-Lys-AMC | 73207-91-7 | 99.8% | 621.68 | 10 MG
SDP
Catalog No. 5000397172
Encompass_Preferred
Suc-Ala-Phe-Lys-AMC is a highly sensitive fluorogenic substrate used for plasmin and assaying gingipain K.
  • High sensitivity for plasmin
  • Suitable for assaying gingipain K
  • White to off-white appearance
  • Powder form
  • Soluble in DMSO
Catalog No. 50-003-97172 Supplier Medchemexpress LLC Supplier No. HYP436010MG
Suc-Ala-Phe-Lys-AMC is a highly sensitive fluorogenic substrate used for plasmin and assaying gingipain K.
  • High sensitivity for plasmin
  • Suitable for assaying gingipain K
  • White to off-white appearance
  • Powder form
  • Soluble in DMSO