Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Suc-Ala-Pro-pNA | 95632-46-5 | 98.5% | 406.39 | 10 MG
SDP

Catalog No. 5000397680
Encompass_Preferred

Suc-Ala-Pro-pNA is a peptide that serves as a substrate to detect the activity and stability of prolyl endopeptidase (PEP). It is a highly pure compound used in research to evaluate the function of prolyl endopeptidase.

  • Substrate for prolyl endopeptidase (PEP)
  • Molecular formula C18H22N4O7
  • Molecular weight 406.39
  • Purity of 98.5%
  • Solid, white to off-white appearance
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-97680 Supplier Medchemexpress LLC Supplier No. HYP449010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suc-Ala-Pro-pNA is a peptide that serves as a substrate to detect the activity and stability of prolyl endopeptidase (PEP). It is a highly pure compound used in research to evaluate the function of prolyl endopeptidase.

  • Substrate for prolyl endopeptidase (PEP)
  • Molecular formula C18H22N4O7
  • Molecular weight 406.39
  • Purity of 98.5%
  • Solid, white to off-white appearance
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.