Learn More
Bachem Suc-Ala-Pro-pNA
SDP
Supplier:  Bachem 4028677.0250
Encompass_Preferred
Suc-Ala-Pro-pNA, 95632-46-5, C18H22N4O7, Mr 406.40, 250mg. Suc-AP-pNA, chromogenic substrate for prolyl endopeptidase.
Catalog No. 50-194-7069