Learn More
Bachem Suc-Ala-Pro-Ala-AMC
SDP
Supplier:  Bachem 4005965.0050
Encompass_Preferred
Suc-Ala-Pro-Ala-AMC, 88467-44-1, C25H30N4O8, Mr 514.54, 50mg. Suc-APA-AMC, a sensitive substrate for human neutrophil elastase.
Catalog No. 50-194-7036