Learn More
Carbosynth LLC. Suc-Glu-Ala-Leu-Phe-Gln-pNA, 25mg, MW: (M.W. 826.87), Molecular formula: C38H50N8O13
SDP
Supplier:  Carbosynth LLC. SAP3693PI25MG
Encompass_Preferred
Synonyms: Succinyl-L-Glutamyl-L-Alanyl-L-Leucyl-L-Phenylalanyl-L-Glutamine p-Nitroanilide, Application: Substrate for 3C Human Rhinovirus-14 (HRV14), Storage Temperature: -20°C, References: Q. M. Wang, R.B. Johnson, G.A. Cox, E.C. Villarreal, and R.J. Loncharich, Anal. Biochem., 252, 238 (1997), Q.M. Wang, R.B. Johnson, L.N. Jungheim, J.D. Cohen, and E.C. Villarreal, Antimicrobial Agents and Chemot., 42, 916 (1998).
Catalog No. 50-168-7036