Learn More
Bachem Suc-Gly-Gly-Phe-pNA
SDP
Supplier:  Bachem 4002298.0250
Encompass_Preferred
Suc-Gly-Gly-Phe-pNA, 68982-90-1, C23H25N5O8, Mr 499.48, 250mg. Suc-GGF-pNA was used to determine the activity of chymotrypsin and of Streptomyces griseus protease B.
Catalog No. 50-260-407