Learn More
Medchemexpress LLC Suc-Gly-Pro-AMC | 80049-85-0 | 99.2% | C21H23N3O7 | 100 MG
SDP
Catalog No. 5000356139
Encompass_Preferred
Suc-Gly-Pro-AMC is a fluorescent and fibroblast activation protein (FAP) specific substrate. It reacts with recombinant porcine prolyl oligopeptidase, making it useful for investigating the activity of FAP and prolyl endopeptidase (PREP). This compound is also utilized in glioma research.
  • Fluorescent substrate
  • Fibroblast activation protein (FAP) specific substrate
  • Reacts with recombinant porcine prolyl oligopeptidase
  • Used to study FAP and prolyl endopeptidase (PREP) activity
  • Utilized in glioma research
Catalog No. 50-003-56139 Supplier Medchemexpress LLC Supplier No. HY145983100MG
Suc-Gly-Pro-AMC is a fluorescent and fibroblast activation protein (FAP) specific substrate. It reacts with recombinant porcine prolyl oligopeptidase, making it useful for investigating the activity of FAP and prolyl endopeptidase (PREP). This compound is also utilized in glioma research.
  • Fluorescent substrate
  • Fibroblast activation protein (FAP) specific substrate
  • Reacts with recombinant porcine prolyl oligopeptidase
  • Used to study FAP and prolyl endopeptidase (PREP) activity
  • Utilized in glioma research