Learn More
Enzo Life Sciences Suc-Leu-Leu-Val-Tyr-AMC (5 mg) CAS: 94367-21-2
SDP
Catalog No. 501031783
Encompass_Preferred
Proteasome substrate. Fluorogenic substrate for 20S proteasome (Prod. No. BML-PW8720) and other chymotrypsin-like proteases, as well as calpains. Ex.: 380nm Em.: 460nm. Alternative Name: Suc-LLVY-AMC, Proteasome substrate (fluorogenic). Calpain substrate (fluorogenic). Formula: C40 H53 N5 O10. MW: 763.9. CAS: 94367-21-2. Formulation: Lyophilized. Purity: ≥97% (HPLC). Sequence N-Succinyl-Leu-Leu-Val-Tyr-AMC (AMC=7-amino-4-methylcoumarin). Solubility: Soluble in DMSO (at least 5mM). Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-103-1783 Supplier Enzo Life Sciences Supplier No. BMLP8020005
Proteasome substrate. Fluorogenic substrate for 20S proteasome (Prod. No. BML-PW8720) and other chymotrypsin-like proteases, as well as calpains. Ex.: 380nm Em.: 460nm. Alternative Name: Suc-LLVY-AMC, Proteasome substrate (fluorogenic). Calpain substrate (fluorogenic). Formula: C40 H53 N5 O10. MW: 763.9. CAS: 94367-21-2. Formulation: Lyophilized. Purity: ≥97% (HPLC). Sequence N-Succinyl-Leu-Leu-Val-Tyr-AMC (AMC=7-amino-4-methylcoumarin). Solubility: Soluble in DMSO (at least 5mM). Long Term Storage: -20°C. Handling: Protect from light.